Multi-layer adhesive floor mats, Ergomat® Sticky Mats
Supplier: ERGOMAT
SM0203
115-2469EA 161 EUR
115-2469
Multi-layer adhesive floor mats, Ergomat® Sticky Mats
Floor Mats Adhesive Floor Mats
Ergomat Sticky Mats provide a fast, convenient, reliable way to clean shoe soles, or other items, before entering a clean and sanitary area. They significantly reduce traffic-borne contaminants, and have proven themselves effective in a wide variety of industries, such as microelectronics, aerospace, pharmaceuticals and food processing. These disposable sticky mats consist of a stack of tear-off polyethylene films (full adhesive sheets) that effectively capture dirt and dust.
  • 30 layers per mat
  • Easily adhere to a frame with double-faced adhesive tape
  • Uses a
More Product Information