Hygienic hand rub, VF+, PURELL®
Supplier: GOJO Industries - Europe Ltd
5896-02-EEU
115-3013EA 94.4 EUR
115-3013
Hygienic hand rub, VF+, PURELL®
Skincare Soaps and Skin Cleansers
PURELL® VF+ Hygienic Hand Rub offers unprecedented sanitising while preserving healthy skin. Product benefits include a short contact time and an exceptionally broad spectrum of activity, which exceeds the European standard for hand sanitisers.
  • Kills 99% of the most common germs which may be harmful
  • Can be used throughout a hospital
PURELL® VF+ Hygienic Hand Rub reaches the highest level of virucidal activity provided by standard EN 14476 + A1. Effective against non-enveloped viruses such as Adenovirus, Rotavirus, and Norovirus in 30 seconds.More Product Information