Potassium iodate 99.4-100.4% ACS

Supplier: Thermo Fisher Scientific
Danger

11602.22 11602.36
11602.22EA 52.8 EUR
11602.22 11602.36
Potassium iodate 99.4-100.4% ACS
Potassium iodate

Purity: 99.4-100.4% CAS No.: 7758-05-6 Molecular Formula: KIO3 Formula Weight: 214.00 Physical Form: Powder Density: 3.89 Melting Point: 560° dec. Solubility: Slightly soluble in boiling water. Insoluble in alcohol Merck Reference: 14,7642 UN No.: 1479 MDL No.: MFCD00011406

Formula: KlO₃
Melting Pt: 560 °C (dec.)
Density: 3,89
Storage Temperature: Ambient
MDL Number: MFCD00011406
CAS Number: 7758-05-6
EINECS: 231-831-9
UN: 1479
ADR: 5.1,II
Merck Index: 14,7642

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR