N,N-Dimethylformamide 99+%, extra pure

Supplier: Thermo Fisher Scientific
Danger

116220010 116220025 116220250 116220050
116220010.EA 60.4 EUR
116220010. 116220025. 116220250. 116220050.
N,N-Dimethylformamide 99+%, extra pure
N,N-Dimethylformamide
Formula: HCON(CH₃)₂
Boiling Pt: 153 °C (1013 hPa)
Melting Pt: –61 °C
Density: 0,945 g/cm³
Flash Pt: 58 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00003284
CAS Number: 68-12-2
EINECS: 200-679-5
UN: 2265
ADR: 3,III
REACH: 01-2119475605-32
Merck Index: 13,03269
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1ac5c5e9758b6c40ccb0cf127b6a6e1c6d608c64&vid=9a5d4e579e195193602c20d3cb39c89ac9c92cbf&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Clear liquid
Water ≤0.2 % (K.F.)
GC ≥99.0 %
Color scale ≤20 APHA
Residue after evaporation ≤0.005 %
Sulfate (SO4) ≤0.002 %
Calcium (Ca) ≤0.001 %
Iron (Fe) ≤2 ppm
Magnesium (Mg) ≤5 ppm
Heavy metals (ICP-OES) ≤5 ppm
Insoluble matter ≤0.005 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR