Barium perchlorate, anhydrous

Supplier: Thermo Fisher Scientific
Danger

11657.22 11657.36
11657.22EA 70.7 EUR
11657.22 11657.36
Barium perchlorate, anhydrous
Barium perchlorate

CAS No.: 13465-95-7 Molecular Formula: Ba(ClO4)2 Formula Weight: 336.24 Physical Form: Powder Density: 3.20 Sensitivity: Hygroscopic Appearance: White Merck Reference: 14,987 UN No.: 1447 MDL No.: MFCD00003440


Desiccant, ribonuclease determination

Formula: Ba(ClO₄)₂
MW: 336.23 g/mol
Melting Pt: 505
Density: 3.20
Flash Pt: 21 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00003440
CAS Number: 13465-95-7
EINECS: 236-710-4
UN: 1447
ADR: 5.1 II
Merck Index: 14,987

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR