Formaldehyde 30% aqueous solution, Q Path®

Supplier: VWR Chemicals
Danger

Synonyms: Paraformaldehyde aqueous solution, Methanal solution, Methylaldehyde solution, Formol, Formaline solution, Formalin, Methylaldehyde

11699031.EA 41.5 EUR
11699031.
Formaldehyde 30% aqueous solution, Q Path®
Formaldehyde

Formalin 30% fixative for tissue samples.


Caution: Store this product at ambient temperature (above 15 °C), because it may polymerise at temperatures below 9 °C. Presence of polymers does not affect product properties. Precipitate can be removed by filtration.

Formula: CH₂O
MW: 30.03 g/mol
Boiling Pt: -19.3
Melting Pt: -92
Storage Temperature: Ambient
MDL Number: MFCD00003274
CAS Number: 50-00-0
EINECS: 200-001-8
UN: 2209
ADR: 8 III
Merck Index: 13,04259

Order Now

Specification Test Results

Assay 28.5 - 31.5 %
Appearance Clear colourless liquid
Density (20/4) 1.055 - 1.085
Ashes Max. 0.08 %
Heavy metals Max. 8 ppm
Formic acid Max. 0.08 %
Methanol 4.0 - 12.5 %
Fe (Iron) Max. 8 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR