Palladium 5% on gamma alumina, powder, reduced

Supplier: Thermo Fisher Scientific

11713.06 11713.14 11713.22
11713.06EA 95.5 EUR
11713.06 11713.14 11713.22
Palladium 5% on gamma alumina, powder, reduced
Palladium

CAS No.: 7440-05-3 Physical Form: Powder Odour: Odourless MDL No.: MFCD03613602

Formula: Pd
MW: 106.420 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00011167
CAS Number: 7440-05-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR