Ruthenium(III) chloride oxide (≥35,0% Ru), ammoniated

Supplier: Thermo Fisher Scientific

Synonyms: Ruthenium oxychloride, Tetraamminechlorohydroxoruthenium(III) chloride, Ruthenium Red

11806.03 11806.06
11806.03EA 103 EUR
11806.03 11806.06
Ruthenium(III) chloride oxide (≥35,0% Ru), ammoniated
Ruthenium(III) chloride oxide

CAS No.: 11103-72-3 Molecular Formula: H42Cl6N14O2Ru3 Formula Weight: 786.35 Physical Form: Crystalline Density: 3.110 Appearance: Red Odour: Odourless Melting Point: 500° dec. Solubility: Soluble in water, ammonia MDL No.: MFCD00011479


Applications: Microscopic stain

Formula: H₄₂Cl₆N₁₄O₂Ru₃
Melting Pt: >500 °C (dec.)
Density: 3,110
Storage Temperature: Ambient
MDL Number: MFCD00011479
CAS Number: 11103-72-3
Merck Index: 14,8300

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR