Ammonium hexachlororhodate(III) hydrate ≥99.99% (metals basis), Premion®

Supplier: Thermo Fisher Scientific
Danger

11812.01 11812.03 11812.06
11812.01EA 0 EUR
11812.01 11812.03 11812.06
Ammonium hexachlororhodate(III) hydrate ≥99.99% (metals basis), Premion®
Ammonium hexachlororhodate (III) monohydrate

Purity: 99.99% CAS No.: 15336-18-2 Molecular Formula: (NH4)3RhCl6.xH2O (x≈1) (formula not proven) Formula Weight: 369.74(anhy) Physical Form: Powder Sensitivity: Hygroscopic Appearance: Dark red Melting Point: dec. MDL No.: MFCD00003394

Formula: H₁₂Cl₆N₃Rh·H₂O
Melting Pt: Dec.
Storage Temperature: Ambient
MDL Number: MFCD00003394
CAS Number: 15336-18-2
EINECS: 239-364-2

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR