Containment berms, QuickBerm®
Supplier: JUSTRITE MANUFACTURING
28512 28514 28516 28518 28519 28522 28524 28528 28532
119-0364EA 718 EUR
119-0364 119-0365 119-0366 119-0367 119-0368 119-0369 119-0370 119-0371 119-0372
Containment berms, QuickBerm®
Spill Control Materials
Ergonomic design provides 100% inside berm accessibility with an unobstructed side-to-side floor space for vehicle tires, foot traffic, and cleaning.
  • Single-piece construction patented Rigid-Lock wall-support system integral to berm provides unobstructed work space
  • Quick and easy setup, no assembly required
  • Durable EnGuard™V, modified PVC coated fabric withstands long-term UV exposure
  • It can withstands a broad range of chemicals including petroleum, oils, grease, and most acids—durable enough for extended wear
  • Helps comply with EPA for containment and
More Product Information