Palladium ≥99.98% (metals basis), foil, Thickness: 0.25 mm

Supplier: Thermo Fisher Scientific

11918.FF 11918.FI
11918.FFEA 725 EUR
11918.FF 11918.FI
Palladium ≥99.98% (metals basis), foil, Thickness: 0.25 mm
Palladium

Purity: 99.98+% CAS No.: 7440-05-3 Physical Form: ≈1.9g/25x25mm Odour: Odourless MDL No.: MFCD00011167

Formula: Pd
MW: 106.420 g/mol
Density: 12.02 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011167
CAS Number: 7440-05-3
EINECS: 231-115-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR