Maintenance sorbents
Supplier: 3M
MF2001 MM1001 MN1001
121-1001EA 612 EUR
121-1001 121-0016 129-0146
Maintenance sorbents
Spill Control Materials
Light grey when dry and darken when wet as an indication of saturation level.
  • Inert, synthetic fibres principally of PP and polyester
  • Multi-format combines four formats in one product and can be used as booms, pillows, sheets or rolls
  • Minibooms are designed for placing around the base of equipment to contain and control leaks and spills
  • Minipillows are high capacity for use at the source of a leak. They are tear resistant and can be left in place for long periods
Used for the control of accidental and routine leaks, drips and spills of oils,More Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable