Copper(I) cyanide

Supplier: Thermo Fisher Scientific
Danger

12135.30 12135.A1 12135.F3
12135.30EA 55.5 EUR
12135.30 12135.A1 12135.F3
Copper(I) cyanide
Copper (I) cyanide

CAS No.: 544-92-3 Molecular Formula: CCuN Formula Weight: 89.56 Physical Form: Powder Density: 2.92 Appearance: White to cream Odour: Characteristic Melting Point: 473° Solubility: Practically insoluble in water, alcohol. Soluble in NH4OH Beilstein Reference: 3587244 Merck Reference: 14,2661 UN No.: 1587 MDL No.: MFCD00010975


Electroplating Cu or Fe, polymerization catalyst

Formula: CCuN
Melting Pt: 473 °C
Density: 2,92
Storage Temperature: Ambient
MDL Number: MFCD00010975
CAS Number: 544-92-3
EINECS: 208-883-6
UN: 1587
ADR: 6.1,II
Merck Index: 14,2661

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR