Gold trihydroxide (≥79% Au)

Supplier: Thermo Fisher Scientific
Warning

12159.03 12159.06 12159.09
12159.03EA 262 EUR
12159.03 12159.06 12159.09
Gold trihydroxide (≥79% Au)
Gold trihydroxide

CAS No.: 1303-52-2 Molecular Formula: Au(OH)3 Formula Weight: 247.99 Physical Form: Powder Sensitivity: Light Sensitive Odour: Odourless Merck Reference: 14,4523 MDL No.: MFCD00046173


Gold plating solutions, porcelain decoration

Formula: H₃AuO₃
Storage Temperature: Ambient
MDL Number: MFCD00046173
CAS Number: 1303-52-2
EINECS: 215-120-0
Merck Index: 14,4523

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR