Potassium hexachloroplatinate(IV) (39,6% Pt)

Supplier: Thermo Fisher Scientific
Danger

12169.06 12169.01 12169.03
12169.06EA 484 EUR
12169.06 12169.01 12169.03
Potassium hexachloroplatinate(IV) (39,6% Pt)
Potassium hexachloroplatinate(IV)

CAS No.: 16921-30-5 Molecular Formula: Cl6K2Pt Formula Weight: 486.01 Physical Form: Crystalline powder Density: 3.50 Refractive Index: ≈1.825 Sensitivity: Hygroscopic Appearance: Yellow Odour: Odourless Melting Point: 250°dec. Solubility: Soluble in hot water. Slightly soluble in cold water. Practically insoluble in alcohol Merck Reference: 14,7636 UN No.: 3288 MDL No.: MFCD00011389

Formula: K₂PtCl₆
Melting Pt: 250 °C, (dec.)
Density: 3,50
Storage Temperature: Ambient
MDL Number: MFCD00011389
CAS Number: 16921-30-5
EINECS: 240-979-3
UN: 3288
ADR: 6.1,III
Merck Index: 14,7636

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR