Cobalt(II) acetate tetrahydrate (24% Co)

Supplier: Thermo Fisher Scientific
Danger

Synonyms: Cobaltous acetate tetrahydrate, Acetic acid cobalt (II) salt tetrahydrate, Cobalt(Ⅱ) acetate tetrahydrate, Cobalt diacetate tetrahydrate

12227.30 12227.A1
12227.30EA 76.1 EUR
12227.30 12227.A1
Cobalt(II) acetate tetrahydrate (24% Co)
Cobalt (II) acetate tetrahydrate

CAS No.: 6147-53-1 Molecular Formula: C4H6CoO4.4H2O Formula Weight: 249.08 (177.02anhy) Physical Form: Crystals Density: 1.71 Refractive Index: 1.542 Sensitivity: Hygroscopic Appearance: Red Odour: Odourless Melting Point: ca 140° -4H2O Solubility: Soluble in water, alcohol, dilute acids Beilstein Reference: 3731397 Merck Reference: 14,2433 UN No.: 3077 MDL No.: MFCD00149569

Formula: (H₃CCOO)₂Co·4H₂O
Melting Pt: ∼140 °C (-4H₂O)
Density: 1,71
Storage Temperature: Ambient
MDL Number: MFCD00149569
CAS Number: 6147-53-1
EINECS: 200-755-8
UN: 3077
ADR: 9,III
Merck Index: 14,2433

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR