Calcium chloride, anhydrous ≥93%

Supplier: Thermo Fisher Scientific
Warning

12316.A1 12316.A7
12316.A1EA 65 EUR
12316.A1 12316.A7
Calcium chloride, anhydrous ≥93%
Calcium chloride

Purity: 93% CAS No.: 10043-52-4 Molecular Formula: CaCl2 Formula Weight: 110.99 Physical Form: Granular powder Sensitivity: Hygroscopic Boiling Point: 1600° Solubility: Freely soluble in water and alcohol Merck Reference: 14,1659 MDL No.: MFCD00010903


As drying and dehydrating agent for organic liquids and gases, and in desiccators

Formula: CaCl₂
Boiling Pt: >1600 °C
Melting Pt: 782 °C
Density: 2,174 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010903
CAS Number: 10043-52-4
EINECS: 233-140-8
REACH: 01-2119494219-28
Merck Index: 14,1659

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR