VWR®, Organic reference standard, Volatile Organic Compound Mix 1 (Liquid)
Supplier: VWR Chemicals
123382JEA 248 EUR
123382J
VWR®, Organic reference standard, Volatile Organic Compound Mix 1 (Liquid)
Standards Organic Standards
0,2 mg/ml of each component in Methanol
Volatile Organic Components by PID/ELCD
54 liquid components :
Benzene; Bromobenzene; Bromochloromethane; Bromodichloromethane; Bromoform; n-Butylbenzene; sec-Butylbenzene; tert-Butylbenzene; Carbon tetrachloride; Chlorobenzene; Chloroform; 2-Chlorotoluene; 4-Chlorotoluene; Dibromochloromethane; 1,2-Dibromo-3-chloropropane; 1,2-Dibromoethane; Dibromomethane; 1,2-Dichlorobenzene; 1,3-Dichlorobenzene; 1,4-Dichlorobenzene; 1,1-Dichloroethane; 1,2-Dichloroethane; 1,1-Dichloroethene; cis-1,2-Dichloroethene; trans-1,2-Dichloroethene; 1,2-Dichloropropane; 1,3-Dichloropropane; 2,2-Dichloropropane; 1,1-Dichloropropene; cis-1,3-Dichloropropene; trans-1,3-Dichloropropene; Ethylbenzene; Hexachlorobutadiene; Isopropylbenzene; p-Isopropyltoluene; Methylene chloride; Naphthalene; n-Propylbenzene; Styrene; 1,1,1,2-Tetrachloroethane; 1,1,1,2-Tetrachloroethane; Tetrachloroethene; Toluene; 1,2,3-Trichlorobenzene; 1,2,4-Trichlorobenzene; 1,1,1-Trichloroethane; 1,1,2-Trichloroethane; Trichloroethene; 1,2,3-Trichloropropane; 1,2,4-Trimethylbenzene; 1,3,5-Trimethylbenzene; o-Xylene; m-Xylene; p-Xylene