Cadmium(II) sulphate 8/3-hydrate 98.0-102.0% ACS

Supplier: Thermo Fisher Scientific
Danger

12340.14 12340.30 12340.A1
12340.14EA 24.6 EUR
12340.14 12340.30 12340.A1
Cadmium(II) sulphate 8/3-hydrate 98.0-102.0% ACS
Cadmium (II) sulphate 8/3-hydrate

Purity: 98.0-102.0% CAS No.: 7790-84-3 Molecular Formula: 3CdSO4.8H2O Formula Weight: 769.50 (208.46anhy) Physical Form: Crystalline Density: 3.08 Refractive Index: 1.565 Appearance: White Odour: Odourless Melting Point: 40° -H2O Solubility: Soluble in water. Almost insoluble in alcohol and ethyl acetate Merck Reference: 14,1627 UN No.: 2570 MDL No.: MFCD00151655


Used in Standard Weston Cell, as electrolytes in electroplating, in phosphors, in manufacture of standard cadmium elements, and as a catalyst in Marsh test for AS

Formula: 3CdSO₄·8H₂O
Melting Pt: >40 °C (-H₂O)
Density: 3,08
Storage Temperature: Ambient
MDL Number: MFCD00151655
CAS Number: 7790-84-3
EINECS: 233-331-6
UN: 2570
ADR: 6.1,III
Merck Index: 14,1627

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR