cis-Dichlorobis(2,2'-bipyridine)ruthenium(II) dihydrate (≥19% Ru)

Supplier: Thermo Fisher Scientific

12778.03 12778.06
12778.03EA 204 EUR
12778.03 12778.06
cis-Dichlorobis(2,2'-bipyridine)ruthenium(II) dihydrate (≥19% Ru)
cis-Dichlorobis(2,2'-bipyridine)ruthenium(II) dihydrate

CAS No.: 15746-57-3 Molecular Formula: C20H16Cl2N4Ru.2H2O Formula Weight: 520.38 (484.35anhy) Physical Form: Crystalline Appearance: Black MDL No.: MFCD00012040

Formula: C₂₀H₂₀Cl₂N₄O₂Ru
Storage Temperature: Ambient
MDL Number: MFCD00015504
CAS Number: 15746-57-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR