Bis(cyclopentadienyl)ruthenium (≥43,2% Ru)

Supplier: Thermo Fisher Scientific
Warning


Packaged under argon.

12781.03 12781.06
12781.03EA 261 EUR
12781.03 12781.06
Bis(cyclopentadienyl)ruthenium (≥43,2% Ru)
Bis(cyclopentadienyl)ruthenium

CAS No.: 1287-13-4 Molecular Formula: C10H10Ru Formula Weight: 231.26 Physical Form: Crystalline, Packaged under argon Sensitivity: Air and Moisture Sensitive Melting Point: 194-198° MDL No.: MFCD00001442

Formula: C₁₀Ru
Melting Pt: 194…198 °C
Storage Temperature: Ambient
MDL Number: MFCD00001442
CAS Number: 1287-13-4
EINECS: 215-065-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR