Diarsenic triselenide ≥99.999% (metals basis)

Supplier: Thermo Fisher Scientific
Danger

12849.02 12849.06 12849.14
12849.02EA 0 EUR
12849.02 12849.06 12849.14
Diarsenic triselenide ≥99.999% (metals basis)
Diarsenic triselenide

Purity: 99.999% CAS No.: 1303-36-2 Molecular Formula: As2Se3 Formula Weight: 386.72 Physical Form: 1-6mm Fused Pieces Density: 4.75 Melting Point: 260° Merck Reference: 14,805 UN No.: 3077 MDL No.: MFCD00014166


For vacuum deposition

Formula: As₂Se₃
Melting Pt: 260 °C
Density: 4,75
Storage Temperature: Ambient
MDL Number: MFCD00014166
CAS Number: 1303-36-2
EINECS: 215-119-5
UN: 3077
ADR: 9,III
Merck Index: 14,805

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR