Cleaning pad, OpticPad®
Supplier: CLEANTEX PRODUCTS
811
129-0150EA 0 EUR
129-0150
Cleaning pad, OpticPad®
Wipes Cleaning Kits and Pads
Ideal for cleaning and polishing front and back surfaces of mirrors, microscope lenses, safety goggles, and photographic equipment.
  • Solvent is filtered to 0,2 µm and evaporates quickly without leaving a residue
  • Disposable, lint-free pad is pre-saturated for cleaning optical grade surfaces
  • Each pad is hermetically sealed and individually wrapped
Packaging: 100 per bag.