MultiSqueegee swivel with mono blade
Supplier: FHP VILEDA SCS
119933
129-0265EA 36.7 EUR
129-0265
MultiSqueegee swivel with mono blade
Mops
For dry or wet push and pull applications. An alternative to a broom in places where bristles are not permitted, e.g. healthcare floors and walls. The MultiSqueegee combines three components: an aluminum rail
, an elastomer rubber blade (washable at 70 °C
) and a universal polypropylene handle connector. The replaceable rubber blade has a single sharp edge that drags the liquid and finest dirt in one go.
  • All materials can be cleaned by washing at 70 °C
  • Components are lightweight and easy to dismantle and the blade can be replaced
  • Materials have been
More Product Information