Bag sealing machines, Superseal Impulse
Supplier: HULME MARTIN HEAT SEALERS
76002051 65002161 30001061 30002161 30202161 25001051 29502051 13001061
129-0394EA 2680 EUR
129-0394 129-0393 129-0388 129-0389 129-0390 129-0386 129-0387 129-0385
Bag sealing machines, Superseal Impulse
Heat Sealers Bag Sealers
Superseal Impulse heat sealers are suitable for a wide range of laboratory and medical applications. Units are used extensively for the protective packaging, storage and distribution of pharmaceutical products, chemicals, and many other materials. They are also used for the safe containment and transfer of blood specimens, medical waste and other potentially hazardous substances.
  • Simple to maintain with consumable spares being easy to fit
  • Seal instantly without warm up delay
Superseal Impulse machines are fitted with 'plug-in' replacement elements forMore Product Information