Microfiber string mops, M-Zone MegaSorb™
Supplier: MICRONOVA
MZCRMS11-12 JZMZCRMS-10 MZCRMS-10IR
129-0619EA 718 EUR
129-0619 MICRJZMZCRMS-10 MICRMZCRMS-10IR
Microfiber string mops, M-Zone MegaSorb™
Mops
A highly absorbent mop that combines the cleaning efficiency of microfibre with the ease of a looped string mop. The tufted strands are comprised of polyester and nylon filaments, designed to pick up liquids and tiny particles in controlled and critical environments.
  • Highly absorbent and low-linting
  • Polyester and nylon tufted microfibre tubes
  • Picks up tiny particles where cotton fibres just roll over
  • White microfibre option includes an additional scrubbing band
  • Use with MH-1 handle or QDMH-1 MicroMop adapter with SSU series handles
  • Compatible with alcohol
More Product Information