VWR®, Autoclavable Bags
Supplier: VWR Collection
129-0018EA 464 EUR
129-0018 129-1153 129-1155 129-1572
VWR®, Autoclavable Bags
Bags Waste Disposal Bags
Sturdy, temperature resistant bags, ideal for disposing of hazardous waste that must first be autoclaved. Can also be used for autoclaving other products. Supplied complete with sealing wires. PP, 37,5 µm or 50 µm thick, transparent.
  • Clear bag
  • Autoclavable up to 135 °C
  • Highly resistant to tears and impact