VWR®, Autoclavable Bags
Supplier: VWR Collection
129-1575EA 827 EUR
129-1575
VWR®, Autoclavable Bags
Bags Waste Disposal Bags
Sturdy, temperature resistant bags, ideal for sterilising animal cages.
  • PE, clear
  • Autoclavable up to 121 °C
  • Highly puncture resistant and impact strength