Metal-in static shielding bags, SCS
Supplier: DESCO
1001012 1001216 1001818 1001824 100810 100812 10035 10046 10058 100610 10068 3001012 3001014 3001418 3001824 30035 30046 30058 300610 30068 300810 1001024 1001224 3001024
129-2609EA 30 EUR
129-2609 129-2610 129-2611 129-2612 129-2613 129-2614 129-2622 129-2623 129-2624 129-2625 129-2626 129-2631 129-2632 129-2633 129-2634 129-2635 129-2636 129-2637 129-2638 129-2639 129-2640 DESC1001024 DESC1001224 DESC3001024
Metal-in static shielding bags, SCS
Bags Storage Bags
Transparent metal-in static shielding bag provides a static safe environment for ESD sensitive electronic devices (ESDS).
  • Printed with an ESD protective symbol and a lot code for traceability
  • Heat sealable or zip-locked
  • Manufactured from a polyester, metal, polyethylene laminate
The polyester dielectric in concert with the metal layer provides discharge shielding. The exterior being static dissipative allows electrostatic charges to be removed when grounded.
Certifications: Complies with IEC 61340-5-1 and IEC 61340-5-3.
Packaging: Suppied perMore Product Information