Bags, self adhesive, Cleanware™, SP Bel-Art
Supplier: Bel-Art Products, a Part of SP
F131741008 F131741012
129-4930EA 52.3 EUR
129-4930 129-4931
Bags, self adhesive, Cleanware™, SP Bel-Art
Bags Waste Disposal Bags
These disposable white waste bags can be hung on free bench or hood space, walls, cabinets or virtually for easy waste collection.
Made of high molecular weight high density (HMHD) polyethylene.
  • Peel backing off the adhesive strip across the top and hang close to your work area
  • Back panel is longer than the front for easy opening
Caution: Maximum weight of the contents should not exceed 1,36 kg.