Dysprosium trinitrate pentahydrate ≥99.9%

Supplier: Thermo Fisher Scientific
Danger

Synonyms: Dysprosium (III) nitrate pentahydrate

12922.14 12922.22
12922.14EA 83.2 EUR
12922.14 12922.22
Dysprosium trinitrate pentahydrate ≥99.9%
Dysprosium trinitrate pentahydrate

Purity: 99.9% CAS No.: 10031-49-9 Molecular Formula: Dy(NO3)3.5H2O Formula Weight: 438.59 (348.51anhy) Physical Form: Crystalline Sensitivity: Hygroscopic Appearance: Cream to pale yellow Odour: Odourless pH: 3 Melting Point: 88.6° Merck Reference: 14,3482 UN No.: 1477 MDL No.: MFCD00149683

Formula: DyN₃O₉·5H₂O
Melting Pt: 88,6 °C
Storage Temperature: Ambient
MDL Number: MFCD00149683
CAS Number: 10031-49-9
EINECS: 233-410-5
UN: 1477
ADR: 5.1,III
Merck Index: 14,3482

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR