Peqlab peqGOLD, Plasmid Purification, Plasmid Miniprep Kit II
Supplier: VWR Chemicals
13-6945-00EA 16 EUR
13-6945-00
Peqlab peqGOLD, Plasmid Purification, Plasmid Miniprep Kit II
Nucleic Acid Reagents Nucleic Acid Purification Kits and Reagents
Isolate up to 40 to 70 µg of high quality plasmid DNA from up to 15 ml bacterial cultures in 30 minutes or less. Simplified with VWR® peqGold Mini Column II technology into three quick steps: Bind, wash and elute.
  • Safe, no phenol/chloroform extractions
  • Versatile with both spin and vacuum formats available
  • High quality DNA is suitable for a wide range of downstream applications
Delivery information: Each kit contains peqGOLD DNA columns, 2 ml collection tubes, Solution I, Solution II, Solution III, HBC buffer, DNA wash buffer, RNase A and elution buffer.