Niobium diselenide ≥99.8% (metals basis)

Supplier: Thermo Fisher Scientific
Danger

Synonyms: Niobium(IV) selenide

13101.09 13101.18
13101.09EA 109 EUR
13101.09 13101.18
Niobium diselenide ≥99.8% (metals basis)
Niobium diselenide

Purity: 99.8% CAS No.: 12034-77-4 Molecular Formula: NbSe2 Formula Weight: 250.83 Physical Form: 5 Micron Powder Odour: Odourless UN No.: 3283 MDL No.: MFCD00016228

Formula: NbSe₂
Melting Pt: >1300 °C
Density: 6,3 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00016228
CAS Number: 12034-77-4
EINECS: 234-811-8
UN: 3283
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR