Ytterbium(III) 2,4-pentanedionate ≥99.9% (REO, rare earth oxide basis)

Supplier: Thermo Fisher Scientific
Warning

Synonyms: Ytterbium(III) acetylacetonate

13159.04 13159.09
13159.04EA 106 EUR
13159.04 13159.09
Ytterbium(III) 2,4-pentanedionate ≥99.9% (REO, rare earth oxide basis)
Ytterbium(III) 2,4-pentanedionate

Purity: 99.9% CAS No.: 14284-98-1 Molecular Formula: C15H21O6Yb Formula Weight: 470.37 Physical Form: Powder Appearance: White Odour: Odourless MDL No.: MFCD00013508

Formula: C₁₅H₂₁O₆Yb
Storage Temperature: Ambient
MDL Number: MFCD00013508
CAS Number: 14284-98-1
EINECS: 238-198-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR