VWR® Mr. Lab, Cleanroom Chairs and Stools
Supplier: VWR Collection
132-0421EA 639 EUR
132-0421 132-0422 132-0503
VWR® Mr. Lab, Cleanroom Chairs and Stools
Chairs
All chairs and stools are available with an aluminium five star base.
  • Gas-lift seat height adjustment
  • Adjustable backrest height and inclination
  • Sealed upholstery, with special foam technology, in artificial leather covering, washable and disinfectant-proof
  • Materials suitable for use in cleanrooms
  • Smooth, sealed surfaces
  • Encapsulated mechanisms
  • Electrostatic discharge measures
Glides: The glides are wear-proof plastic, and have a large surface area, offering a high level of stability.
Castors: Dual-wheel castors, with load-sensitive brakes, have toMore Product Information