Laminar flow workstations, A range
Supplier: BASSAIRE
A2HB A2HF A2VB A2VF A3HB A3HF A3VB A3VF A4HB A4HF A4VB A4VF A5HB A5HF A5VB A5VF A6HB A6HF A6VB
135-0000EA 0 EUR
135-0000 135-0001 135-0002 135-0003 135-0004 135-0005 135-0006 135-0007 135-0008 135-0009 135-0010 135-0011 135-0012 135-0013 135-0014 135-0015 135-0016 135-0017 135-0018
Laminar flow workstations, A range
Fume Hoods and Enclosures Laminar Flow Cabinets
These laminar flow workstations have been designed to protect product and operating staff by maintaining a dust free ultra clean environment. The A-range offers a more superior design being of one-piece laminated construction with a laminated post formed worktop. This design eliminates the use of aluminium tubular section thereby offering improved rigidity.
  • Combine safety and reliability
  • User-friendly and easy to clean and sanitize
  • Provide an aesthetic appearance
  • Choice of five sizes in horizontal (H) or vertical (V) flow configuration
  • Bench (B) or free
More Product Information