Laminar flow workstations, K range
Supplier: BASSAIRE
K2 K3 K4 K5 K6 A6VF
135-0020EA 0 EUR
135-0020 135-0021 135-0022 135-0023 135-0024 135-0019
Laminar flow workstations, K range
Fume Hoods and Enclosures Laminar Flow Cabinets
Designed to protect product and operating staff by maintaining a dust free ultra clean environment. The K-range is constructed from anodised aluminium tubular section with clear acrylic infill panels. K-range enclosures are available assembled or in kit form if required (please specify).
  • Combine safety and reliability
  • User-friendly and easy to clean and sanitise
  • Choice of five sizes in horizontal (H) or vertical (V) flow configuration
  • Bench (B) or free standing (F) models
  • 13 amp socket outlet and variable speed controllers
  • Easy self-assemble, particle-free
More Product Information