Laminar flow workstations, P range
Supplier: BASSAIRE
P2HB P2HF P2VB P2VF P3HB P3HF P3VB P3VF P4HB P4HF P4VB P4VF P5HB P5HF P5VB P5VF P6HB P6HF P6VB P6VF
135-0026EA 0 EUR
135-0026 135-0027 135-0028 135-0029 135-0030 135-0031 135-0032 135-0033 135-0034 135-0035 135-0036 135-0037 135-0038 135-0039 135-0040 135-0041 135-0042 135-0025 135-0043 135-0044
Laminar flow workstations, P range
Fume Hoods and Enclosures Laminar Flow Cabinets
These laminar flow workstations have been designed to protect product and operating staff by maintaining a dust free ultra clean environment. The P-range is constructed in white powder coated high quality carbon steel with a 304 L stainless steel post formed worktop.
  • Combine safety and reliability
  • User-friendly and easy to clean and sanitize
  • Provide an aesthetic appearance
  • Choice of five sizes in horizontal (H) or vertical (V) flow configuration
  • Bench (B) or free standing (F) models
  • 13 amp socket outlet and variable speed controllers
A variety ofMore Product Information