Biological safety cabinet, BioVanguard B, for cytotoxic (high risk) applications (cytotoxic medicines)

Supplier: CleanAir by Baker
NH611003 NH411003 NH411001 NH611001
135-1871EA 22560 EUR
135-1871 135-1867 135-1865 135-1869
Biological safety cabinet, BioVanguard B, for cytotoxic (high risk) applications (cytotoxic medicines)
Fume Hoods and Enclosures Biological Safety Cabinets
BioVanguard Greenline B is a class II cabinet range which offers simplicity, robustness and high reliability ensuring the highest level of protection for operator, product and the environment, minimising hazards inherent to working with agents assigned to biosafety levels 1, 2 and 3. BioVanguard B is designed for high risk microbiological and high toxic applications, such as the production of cytotoxic medicines.

  • Easy to install, easy to operate, easy to clean, easy to decontaminate with low maintenance
  • Microprocessor control with automatic filter clogging
More Product Information
Order Now

SPECIFICATIONS

Model BioVanguard 4 B BioVanguard 6 B
Environmentally Preferable
Filters Pre-filter (EN 779): G3
Downflow HEPA filter (EN 1822): H14
Exhaust HEPA filter (EN 1822): H14
1 st HEPA filter (EN 1822)
Pre-filter (EN 779): G3
Downflow HEPA filter (EN 1822): H14
Exhaust HEPA filter (EN 1822): H14
1 st HEPA filter (EN 1822)
Average air velocity 0,36 m/s 0,36 m/s
Lighting system 1150 lux 1350 lux
Noise level 54 dB(A) 58 dB(A)
Electrical connection 230 V, 50 Hz 230 V, 50 Hz
Power rating 0,196 kW 0,340 kW
Ext. W×D×H 1374×794×1602 mm 1984×794×1602 mm
Int. W×D×H 1180×550×744 mm 1790×550×744 mm
Weight 270 kg 400 kg

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR