MSC-Advantage™ Microbiological Safety Cabinet, Class II
Supplier: Thermo Scientific
51025411 51025413 51028225 51028226
135-6834EA 12770 EUR
135-6834 135-6835 135-1327 135-1328
MSC-Advantage™ Microbiological Safety Cabinet, Class II
Fume Hoods and Enclosures Biological Safety Cabinets
This biological safety cabinet combines smart design and extraordinary value with best in class energy efficiency, reliability and usability. The advanced design of the MSC-Advantage biological safety cabinet improves overall operation and maintenance, maximises safety and optimises service and clean-up. With advanced internal airflow and motor technology, MSC-Advantage cabinets offer optimal energy efficiency, operation and serviceability. At full speed the motors consume 60% less energy than standard cabinets. To further conserve energy, the intelligent speed control automatically reducesMore Product Information