Silica gel 2-5 mm drying agent heavy metal-free, with moisture indicator (orange gel), Fluka™

Supplier: Honeywell Chemicals

13767-1KG 13767-2.5KG 13767-500G 13767-6X1KG 13767-6X500G 13767-6X2.5KG
13767-1KGEA 119 EUR
13767-1KG 13767-2.5KG 13767-500G 13767-6X1KG 13767-6X500G 13767-6X2.5KG
Silica gel 2-5 mm drying agent heavy metal-free, with moisture indicator (orange gel), Fluka™
Silica gel

Silica gel with moisture indicator free of heavy metals.

Formula: SiO₂
Boiling Pt: 2,230 °C
Melting Pt: 1,600 °C
Density: 2 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011232
CAS Number: 112926-00-8
EINECS: 231-545-4

Order Now

Specification Test Results

Soluble in chloroform Max. 0.025 %
Water absorbing capacity Min. 25 %
Loss on drying (150°C, 3h) Max. 2 %
Capacity ≥25 % absorption capacity (water)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR