N,N-Dimethylformamide ≥99.7% for spectrophotometry

Supplier: Thermo Fisher Scientific
Danger

13808.K2 13808.K7 13808.M1
13808.K2EA 89.1 EUR
13808.K2 13808.K7 13808.M1
N,N-Dimethylformamide ≥99.7% for spectrophotometry
N,N-Dimethylformamide

Purity: 99.7+% CAS No.: 68-12-2 Molecular Formula: HCON(CH3)2 Formula Weight: 73.09 Flash Point: 57°(135°F) Physical Form: Liquid Density: 0.944 Refractive Index: 1.4310 Odour: Amine-like pH: 7 Melting Point: -61° Boiling Point: 153° Beilstein Reference: 605365 Merck Reference: 14,3243 UN No.: 2265 MDL No.: MFCD00003284

Formula: HCON(CH₃)₂
Boiling Pt: 153 °C
Melting Pt: –61 °C
Density: 0,944
Flash Pt: 57 °C (135 °F)
Storage Temperature: Ambient
MDL Number: MFCD00003284
CAS Number: 68-12-2
EINECS: 200-679-5
UN: 2265
ADR: 3,III
REACH: 01-2119475605-32
Merck Index: 14,3243
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1ac5c5e9758b6c40ccb0cf127b6a6e1c6d608c64&vid=9a5d4e579e195193602c20d3cb39c89ac9c92cbf&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR