Nickel standard solution, 1000 mg/L Ni in 5% nitric acid (from Ni), standard for ICP, Specpure®

Supplier: Thermo Fisher Scientific

13839.AD 13839.AE 13839.AP
13839.ADEA 62.9 EUR
13839.AD 13839.AE 13839.AP
Nickel standard solution, 1000 mg/L Ni in 5% nitric acid (from Ni), standard for ICP, Specpure®
Standards Cationic Standards Nickel Standards

Molecular Formula: Ni in 5% HNO₃
Physical Form: Liquid
UN No.: 3264
MDL No.: MFCD00011137


Certifications: Traceable to NIST

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR