Tantalum standard solution, 1000 mg/L Ta in 2% hydrofluoric acid (from TaCl₅), standard for ICP, Specpure®

Supplier: Thermo Fisher Scientific

13840.AD 13840.AE 13840.AP
13840.ADEA 81.2 EUR
13840.AD 13840.AE 13840.AP
Tantalum standard solution, 1000 mg/L Ta in 2% hydrofluoric acid (from TaCl₅), standard for ICP, Specpure®
Standards Cationic Standards Tantalum Standards

Molecular Formula: TaCl₅ in 2% HF
Physical Form: Liquid
UN No.: 1790
MDL No.: MFCD00011252


Certifications: Traceable to NIST

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR