Copper standard solution, 1000 mg/L Cu in 5% nitric acid (from Cu), standard for ICP, Specpure®

Supplier: Thermo Fisher Scientific

13867.AD 13867.AE 13867.AP
13867.ADEA 62.9 EUR
13867.AD 13867.AE 13867.AP
Copper standard solution, 1000 mg/L Cu in 5% nitric acid (from Cu), standard for ICP, Specpure®
Standards Cationic Standards Copper Standards

Molecular Formula: Cu in 5% HNO₃
Physical Form: Liquid
UN No.: 3264
MDL No.: MFCD00010965


Certifications: Traceable to NIST

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR