Laboratory and glassware washers, PG 8535 and PG 8536
Supplier: MIELE
7339400 7339410 7339390 7339380 7339370 7339360 7321240 7870410 7870440 9062950 7831480 7870450 7831710 9062920 7831680 7831500 7831490 7870430 7870460 7907620 9358490 7870470 7870420 9062900
141-0569EA 0 EUR
141-0569 141-0570 141-0568 141-0567 141-0566 141-0565 141-0560 141-0603 141-0606 141-0638 141-0590 141-0607 141-0597 141-0635 141-0595 141-0592 141-0591 141-0605 141-0608 141-0612 141-0644 141-0609 141-0604 141-0633
Laboratory and glassware washers, PG 8535 and PG 8536
Glassware Washing Machines
These cleaning and disinfection machines with integrated drying system and drawer have been designed for the treatment of clinical instruments and for washing laboratory glassware in research laboratories, biology and pharmaceutical industries. The new PG 85 range offers a wide range of innovations such as freely programmable PROFITRONIC+ controls, PerfectSpeed spray arm monitoring, temperature independent PerfectFlow dispensing control on the basis of ultrasound as well as PerfectPureSensor conductivity metering.
  • Freely programmable PROFITRONIC+ controls with 64 program slots and
More Product Information