Isolator cleaning tool
Supplier: Texwipe
STX7114 TX7105 TX7120 TX7121 TX7122 STX7114M
141-1046EA 1010 EUR
141-1046 141-1054 141-1061 141-1062 141-1063 141-1047
Isolator cleaning tool
Mops
Isolator cleaning tool: Mop hardware, replacement head, replacement handle.
Specially engineered to work with isolators and other small areas, this easy to manoeuvre tool offers lightweight, one handed operation. With a low profile, swivel head, the isolator cleaning tool gets into 'hard to reach' places.
Engineered for quick cover changes during operation, the padded head allows for a consistent mop across different surfaces without losing cleaning power. The rounded edges clean the isolator corners and prevent scratches or damage to surfaces.
  • Rapid change system allows user to replace
More Product Information