Descalers, SURE®
Supplier: Diversey
100892014 100892015
141-1584EA 62.2 EUR
141-1584 141-1585
Descalers, SURE®
Cleaning Agents Detergents and Surface Cleansers
Concentrated acidic cleaner for removal of lime scale from kitchen equipment and utensils.
  • Specially developed for frequent descaling
  • Excellent adherence on vertical surfaces
  • Very efficient on micro incrustations and mineral soiling
  • No fragrance or dye
  • Contains plant-based ingredients from renewable sources
  • Decomposed by natural processes
  • No environmental hazard classifications, no safety warnings for product in use for user
SURE® descaler is part of a unique range of plant-based, cleaning products that deliver the professional cleaning results youMore Product Information