Lead(II) oxide 99,99 Metals basis

Supplier: Thermo Fisher Scientific
Danger

Synonyms: Lead monoxide

14240.18 14240.30 14240.A1
14240.18EA 75.3 EUR
14240.18 14240.30 14240.A1
Lead(II) oxide 99,99 Metals basis
Lead (II) oxide

Purity: 99.99% CAS No.: 1317-36-8 Molecular Formula: OPb Formula Weight: 223.19 Physical Form: Powder Density: 9.53 Appearance: Yellow Odour: Odourless Melting Point: 886° Merck Reference: 14,5413 UN No.: 3077 MDL No.: MFCD00011164

Formula: PbO
MW: 223.2 g/mol
Boiling Pt: 1470
Melting Pt: 886 °C
Density: 9.53
Storage Temperature: Ambient
MDL Number: MFCD00011164
CAS Number: 1317-36-8
EINECS: 215-267-0
UN: 3077
ADR: 9 III
Merck Index: 14,5413

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR