Silver 99,9 Metals basis, tubing 6.35 mm (0.25 in) OD, 4.35 mm (0.171 in) ID

Supplier: Thermo Fisher Scientific

14259.BSEA 109 EUR
14259.BS 14259.BW 14259.VC
Silver 99,9 Metals basis, tubing 6.35 mm (0.25 in) OD, 4.35 mm (0.171 in) ID
Silver

Purity: 99.9% CAS No.: 7440-22-4 Physical Form: ≈18g/10cm, Temper: as drawn Odour: Characteristic MDL No.: MFCD00003397

Formula: Ag
MW: 107.87 g/mol
Boiling Pt: 2212
Melting Pt: 962
Density: 10.5 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003397
CAS Number: 7440-22-4
EINECS: 231-131-3
Merck Index: 12,08647

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR